Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADCYAP1, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89120257 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-120-257 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-120-257 Supplier Abnova Corporation Supplier No. PAB18702
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of ADCYAP1.

This gene encodes adenylate cyclase activating polypeptide 1. Mediated by adenylate cyclase activating polypeptide 1 receptors, this polypeptide stimulates adenylate cyclase and subsequently increases the cAMP level in target cells. Adenylate cyclase activating polypeptide 1 is not only a hypophysiotropic hormone, but also functions as a neurotransmitter and neuromodulator. In addition, it plays a role in paracrine and autocrine regulation of certain types of cells. This gene encodes three different mature peptides, including two isotypes, a shorter form and a longer form. Two transcript variants encoding the same protein have been found for this gene. [provided by RefSeq

Sequence: C-GKRYKQRVKNKGRR

Specifications

Antigen ADCYAP1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of ADCYAP1.
Dilution ELISA (1:2000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA)
Gene ADCYAP1
Gene Accession No. NP_001108.2
Gene Alias MGC126852/PACAP
Gene Symbols ADCYAP1
Host Species Goat
Immunogen A synthetic peptide corresponding to amino acids at C-terminus of human ADCYAP1.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116
Test Specificity Reported variants represent identical protein: NP_001093203.1, NP_001108.2.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.