Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-AGER, Polyclonal, Abnova™

Catalog No. 89137688 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-688 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-688 Supplier Abnova Corporation Supplier No. PAB13812

Specifications

Antigen AGER
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Formulation In buffer containing 0.09% sodium azide
Host Species Goat
Immunogen A synthetic peptide corresponding to amino acids 385-399 of AGER.
Quantity 200 μL
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.