Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AGRP, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89132676 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-132-676 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-132-676 Supplier Abnova Corporation Supplier No. PAB6775
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of AGRP.

Agouti-related protein is an antagonist of the melanocortin-3 and melanocortin-4 receptor. It appears to regulate hypothalamic control of feeding behavior via melanocortin receptor and/or intracellular calcium regulation. Agouti-related protein is alternatively spliced into 2 variants which differ in 5' untranslated sequence length. [provided by RefSeq]

Sequence: CRKLGTAMNPCSRT

Specifications

Antigen AGRP
Applications ELISA, Immunohistochemistry (Frozen)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of AGRP.
Dilution ELISA (1:32000) Immunohistochemistry (Frozen sections) (0.05-0.1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 0.5mg/mL Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA)
Gene AGRP
Gene Accession No. NP_001129.1;NP_015531.1
Gene Alias AGRT/ART/ASIP2/MGC118963
Gene Symbols AGRP
Host Species Goat
Immunogen A synthetic peptide corresponding to C-terminus of human AGRP.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 181
Test Specificity This antibody is expected to recognize both reported isoforms (as represented by NP_001129, NP_015531).
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.