Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-AKT1, Polyclonal, Abnova™

Catalog No. 89126303 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-126-303 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-126-303 Supplier Abnova Corporation Supplier No. PAB24586

Specifications

Antigen AKT1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation In 0.5mg/mL Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA)
Gene Accession No. NP_005154.2;NP_001014431.1;NP_001014432.1
Gene Symbols AKT1
Host Species Goat
Immunogen A synthetic peptide corresponding to internal region of human AKT1.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 207
Target Species Human
Content And Storage Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.