Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALDOA Goat anti-Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89115035 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-035 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-035 Supplier Abnova Corporation Supplier No. PAB11351
Add to Cart
Add to Cart

Goat polyclonal antibody raised against native ALDOA.

Specifications

Antigen ALDOA
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Goat polyclonal antibody raised against native ALDOA.
Dilution Sandwich ELISA (1:10000-1:40000) Immunoprecipitation (5 ul) The optimal working dilution should be determined by the end user.
Formulation In 20mM potassium phosphate buffer, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gene ALDOA
Gene Symbols ALDOA
Host Species Goat
Immunogen Native purified ALDOA from rabbit muscle.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100009055
Target Species Rabbit
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.