Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALMS1, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89132348 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-132-348 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-132-348 Supplier Abnova Corporation Supplier No. PAB6354
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of ALMS1.

This gene encodes a protein containing a large tandem-repeat domain. The mouse ortholog of this gene has been shown to function in ciliogenesis in inner medullary collecting duct cells. Mutations in this gene have been associated with Alstrom syndrome. Alternative splice variants have been described but their full length sequences have not been determined

Sequence: C-RVTNQLLGRKVPWD

Specifications

Antigen ALMS1
Applications ELISA, Immunohistochemistry (PFA fixed)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of ALMS1.
Dilution ELISA (1:32000) Immunohistochemistry (1-3 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene ALMS1
Gene Accession No. NP_055935.3
Gene Alias ALSS/DKFZp686A118/DKFZp686D1828/KIAA0328
Gene Symbols ALMS1
Host Species Goat
Immunogen A synthetic peptide corresponding to human ALMS1.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7840
Test Specificity The C terminus of ALMS1 shares an 8 amino acid stretch with two other human proteins: hypothetical protein (XP_169104) and ERAL1 (NP_005693). However, this stretch is located internally in these two proteins and so will most likely not cross-react with this antibody. The N terminus was not chosen as the peptide is expected to cyclise during synthesis.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.