Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANLN Goat anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89132436 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-132-436 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-132-436 Supplier Abnova Corporation Supplier No. PAB6482
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of ANLN.

Sequence: WQPDACYKPIGKP

Specifications

Antigen ANLN
Applications ELISA, Immunohistochemistry (PFA fixed)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of ANLN.
Dilution ELISA (1:64000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (3-10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene ANLN
Gene Accession No. NP_061155.2
Gene Alias DKFZp779A055/Scraps/scra
Gene Symbols ANLN
Host Species Goat
Immunogen A synthetic peptide corresponding to human ANLN.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54443
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.