Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APH1A, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89133128 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-133-128 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-133-128 Supplier Abnova Corporation Supplier No. PAB7304
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of APH1A.

APH1 is a multipass transmembrane protein that interacts with presenilin (see PSEN1; MIM 104311) and nicastrin (APH2; MIM 605254) as a functional component of the gamma-secretase complex. The gamma-secretase complex is required for the intramembrane proteolysis of a number of membrane proteins, including the amyloid-beta precursor protein (APP; MIM 104760) and Notch (MIM 190198).[supplied by OMIM

Sequence: C-HVTDRSDARLQYG

Specifications

Antigen APH1A
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of APH1A.
Dilution ELISA (1:128000) Western Blot (0.05-0.1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene APH1A
Gene Accession No. NP_001071096.1;NP_057106.2
Gene Alias 6530402N02Rik/APH-1A/CGI-78
Gene Symbols APH1A
Host Species Goat
Immunogen A synthetic peptide corresponding to human APH1A.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51107
Test Specificity This antibody is expected to recognize both reported isoforms (NP_001071096.1 and NP_057106.2).
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.