Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Apoa1, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89116867 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-867 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-867 Supplier Abnova Corporation Supplier No. PAB13882
Add to Cart
Add to Cart

Goat polyclonal antibody raised against native Apoa1.

Specifications

Antigen Apoa1
Applications Radioimmune Assays (RIA)
Classification Polyclonal
Conjugate Unconjugated
Description Goat polyclonal antibody raised against native Apoa1.
Dilution The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.09% sodium azide, 100U/mL Penicillin, 10μg/mL Streptomycin, 25ng/mL Amphotericin B.
Gene Apoa1
Gene Alias Alp-1/Apoa-1/Brp-14/Ltw-1/Lvtw-1/MGC102525/Sep-1/Sep-2/Sep2
Gene Symbols Apoa1
Host Species Goat
Immunogen Native purified Apoa1 from mouse plasma high density lipoprotein.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11806
Test Specificity Recognizes mouse and rat Apo AI. Does not react well with human Apo AI.
Target Species Mouse, Rat
Content And Storage Store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.