Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APOL6 Goat anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89117678 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-678 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-678 Supplier Abnova Corporation Supplier No. PAB15606
Add to Cart
Add to Cart

Goat polyclonal antibody raised aganist synthetic peptide of APOL6.

This gene is a member of the apolipoprotein L gene family. The encoded protein is found in the cytoplasm, where it may affect the movement of lipids or allow the binding of lipids to organelles. [provided by RefSeq

Sequence: QRDEDDAPLCED

Specifications

Antigen APOL6
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised aganist synthetic peptide of APOL6.
Dilution ELISA (1:8000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene APOL6
Gene Accession No. NP_085144.1
Gene Alias APOL-VI/APOLVI/DKFZp667M075/FLJ38562/FLJ90164/MGC57495
Gene Symbols APOL6
Host Species Goat
Immunogen A synthetic peptide corresponding to human APOL6.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80830
Test Specificity Approximately 38 KDa band observed in human kidney lysates (calculated MW of 38.1 KDa according to NP_085144.1).
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.