Learn More
ARL4D, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of ARL4D.
Supplier: Abnova Corporation PAB19642
Description
ADP-ribosylation factor 4D is a member of the ADP-ribosylation factor family of GTP-binding proteins. ARL4D is closely similar to ARL4A and ARL4C and each has a nuclear localization signal and an unusually high guanine nucleotide exchange rate. This protein may play a role in membrane-associated intracellular trafficking. Mutations in this gene have been associated with Bardet-Biedl syndrome (BBS). [provided by RefSeq
Sequence: C-KQDQPGALSAAEVEKRSpecifications
ARL4D | |
Polyclonal | |
Goat polyclonal antibody raised against synthetic peptide of ARL4D. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
NP_001652.2 | |
ARL4D | |
A synthetic peptide corresponding to internal region of human ARL4D. | |
100 μg | |
Primary | |
This antibody is expected not to cross-react to other ARL4 products. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
Unconjugated | |
ELISA (1:64000) Western Blot (0.3-1 ug/mL) The optimal working dilution should be determined by the end user. | |
ARL4D | |
ARF4L/ARL6 | |
Goat | |
Antigen affinity purification | |
RUO | |
379 | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only