Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARR3, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89117748 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-748 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-748 Supplier Abnova Corporation Supplier No. PAB15683
Add to Cart
Add to Cart

Goat polyclonal antibody raised aganist synthetic peptide of ARR3.

Sequence: C-RKGEEESQKAVE

Specifications

Antigen ARR3
Applications ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised aganist synthetic peptide of ARR3.
Dilution ELISA (1:1000) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene ARR3
Gene Accession No. NP_004303.1
Gene Alias ARRX
Gene Symbols ARR3
Host Species Goat
Immunogen A synthetic peptide corresponding to human ARR3.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 407
Test Specificity Not yet tested due to unavailability of retinal tissue. At this stage we are dependent on researchers in the field for further characterization of this product.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.