Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATG16L1, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89115134 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-134 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-134 Supplier Abnova Corporation Supplier No. PAB11500
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of ATG16L1.

Autophagy is the major intracellular degradation system delivering cytoplasmic components to lysosomes, and it accounts for degradation of most long-lived proteins and some organelles. Cytoplasmic constituents, including organelles, are sequestered into double-membraned autophagosomes, which subsequently fuse with lysosomes. ATG16L1 is a component of a large protein complex essential for autophagy (Mizushima et al., 2003 [PubMed 12665549]).[supplied by OMIM

Sequence: C-KVEKVLSKQHSSSIN

Specifications

Antigen ATG16L1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of ATG16L1.
Dilution ELISA (1:32000) Western Blot (0.01-0.1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene ATG16L1
Gene Accession No. NP_110430.5;NP_060444.3
Gene Alias APG16L/ATG16L/FLJ00045/FLJ10035/FLJ10828/FLJ22677/IBD10/WDR30
Gene Symbols ATG16L1
Host Species Goat
Immunogen A synthetic peptide corresponding near C-terminus of human ATG16L1.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55054
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.