Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

atoh7, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89115135 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-135 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-135 Supplier Abnova Corporation Supplier No. PAB11501
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of atoh7.

Sequence: C-QFDGLQTEGYFMHYD

Specifications

Antigen atoh7
Applications ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of atoh7.
Dilution ELISA (1:32000) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene atoh7
Gene Accession No. NP_571707.1
Gene Alias ath5/zath5
Gene Symbols atoh7
Host Species Goat
Immunogen A synthetic peptide corresponding to zebrafish atoh7.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58216
Target Species Zebrafish
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.