Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Avian Influenza Hemagglutinin 2, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89137571 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-571 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-571 Supplier Abnova Corporation Supplier No. PAB12965
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of Avian Influenza Hemagglutinin 2.

Specifications

Antigen Avian Influenza Hemagglutinin 2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of Avian Influenza Hemagglutinin 2.
Dilution ELISA (1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene Accession No. AAT76166
Host Species Goat
Immunogen A synthetic peptide corresponding to N-terminus 12 amino acids of Avian Influenza Hemagglutinin 2.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Influenza virus
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.