Learn More
B3GNT2, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of B3GNT2.
Supplier: Abnova Corporation PAB7522
Description
This gene encodes a member of the beta-1,3-N-acetylglucosaminyltransferase family. This enzyme is a type II transmembrane protein. It prefers the substrate of lacto-N-neotetraose, and is involved in the biosynthesis of poly-N-acetyllactosamine chains. [provided by RefSeq
Sequence: C-EKHKGFRTFDIESpecifications
B3GNT2 | |
Polyclonal | |
Unconjugated | |
ELISA (32000) Western Blot (0.1-0.3 ug/mL) The optimal working dilution should be determined by the end user. | |
B3GNT2 | |
B3GN-T1/B3GN-T2/B3GNT/B3GNT-2/B3GNT1/BETA3GNT | |
Goat | |
Antigen affinity purification | |
RUO | |
10678 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of B3GNT2. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
NP_006568.2 | |
B3GNT2 | |
A synthetic peptide corresponding to human B3GNT2. | |
100 μg | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only