Learn More
BAIAP2 Goat anti-Human, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of BAIAP2.
Supplier: Abnova Corporation PAB6057
Description
The protein encoded by this gene has been identified as a brain-specific angiogenesis inhibitor (BAI1)-binding protein. This adaptor protein links membrane bound G-proteins to cytoplasmic effector proteins. This protein functions as an insulin receptor tyrosine kinase substrate and suggests a role for insulin in the central nervous system. It also associates with a downstream effector of Rho small G proteins, which is associated with the formation of stress fibers and cytokinesis. This protein is involved in lamellipodia and filopodia formation in motile cells and may affect neuronal growth-cone guidance. This protein has also been identified as interacting with the dentatorubral-pallidoluysian atrophy gene, which is associated with an autosomal dominant neurodegenerative disease. Alternative splicing results in multiple transcript variants encoding distinct isoforms
Sequence: C-SGSGTLVSTVSpecifications
BAIAP2 | |
Polyclonal | |
Unconjugated | |
ELISA (1:64000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user. | |
BAIAP2 | |
BAP2/IRSP53 | |
Goat | |
Antigen affinity purification | |
RUO | |
10458 | |
Human | |
Liquid |
ELISA, Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of BAIAP2. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
NP_059344.1 | |
BAIAP2 | |
A synthetic peptide corresponding to human BAIAP2. | |
100 μg | |
Primary | |
This antibody is expected to recognize only human isoform 1 according to NP_059344.1. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only