Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCOR, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89132406 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-132-406 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-132-406 Supplier Abnova Corporation Supplier No. PAB6440
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of BCOR.

The protein encoded by this gene was identified as an interacting corepressor of BCL6, a POZ/zinc finger transcription repressor that is required for germinal center formation and may influence apoptosis. This protein selectively interacts with the POZ domain of BCL6, but not with eight other POZ proteins. Specific class I and II histone deacetylases (HDACs) have been shown to interact with this protein, which suggests a possible link between the two classes of HDACs. Several transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: LSATPLYGNVHSW-C

Specifications

Antigen BCOR
Applications ChIP Assay, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of BCOR.
Dilution ELISA (1:2000) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene BCOR
Gene Accession No. NP_060215;NP_065977
Gene Alias ANOP2/FLJ20285/FLJ38041/KIAA1575/MAA2/MCOPS2/MGC131961/MGC71031
Gene Symbols BCOR
Host Species Goat
Immunogen A synthetic peptide corresponding to human BCOR.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54880
Test Specificity This antibody is expected to recognize both reported isoforms (represented by NP_060215 and NP_065977).
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.