Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMI1, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89134546 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-134-546 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-134-546 Supplier Abnova Corporation Supplier No. PAB9982
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of BMI1.

Specifications

Antigen BMI1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of BMI1.
Dilution ELISA (1:5000-1:30000) Western Blot (1:500-1:3000) The optimal working dilution should be determined by the end user.
Formulation In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gene BMI1
Gene Accession No. NP_005171;P35226
Gene Alias MGC12685/PCGF4/RNF51
Gene Symbols BMI1
Host Species Goat
Immunogen A synthetic peptide corresponding to amino acids 252-264 of human BMI1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 648
Target Species Bovine, Canine, Chicken, Chimpanzee, Feline, Frog, Human, Orangutan, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.