Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C10orf59 Goat anti-Human, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89132686 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-132-686 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-132-686 Supplier Abnova Corporation Supplier No. PAB6786
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of C10orf59.

Renalase is a flavin adenine dinucleotide-dependent amine oxidase that is secreted into the blood from the kidney (Xu et al., 2005 [PubMed 15841207]).[supplied by OMIM

Sequence: C-VSIDNKKRNI

Specifications

Antigen C10orf59
Applications ELISA, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of C10orf59.
Dilution ELISA (1:32000) Western Blot (0.1-0.3 ug/mL) Immunohistochemistry (3 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene C10orf59
Gene Accession No. NP_001026879.1;NP_060833.1
Gene Alias FLJ11218/RENALASE
Gene Symbols C10orf59
Host Species Goat
Immunogen A synthetic peptide corresponding to amino acids 224-233 of human C10orf59.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55328
Test Specificity This antibody is expected to recognize both reported isoforms (NP_001026879.1and NP_060833.1).
Target Species Human, Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.