Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C13orf3, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89118908 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-118-908 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-118-908 Supplier Abnova Corporation Supplier No. PAB17243
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of C13orf3.

This gene encodes a component of the spindle and kinetochore-associated protein complex that regulates microtubule attachment to the kinetochores during mitosis. The encoded protein localizes to the outer kinetechore and may be required for normal chromosome segregation and cell division. Alternative splicing results in multiple transcript variants. [provided by RefSeq

Sequence: C-PLSKTNSSSNDLE

Specifications

Antigen C13orf3
Applications ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of C13orf3.
Dilution ELISA (1:64000) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene C13orf3
Gene Accession No. NP_659498.3
Gene Alias MGC4832/RAMA1
Gene Symbols C13orf3
Host Species Goat
Immunogen A synthetic peptide corresponding to amino acids at internal region of human C13orf3.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221150
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.