Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C3 Goat anti-Rat, Polyclonal Antibody, Abnova™

Catalog No. 89117593 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-593 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-593 Supplier Abnova Corporation Supplier No. PAB15380
Add to Cart
Add to Cart

Goat polyclonal antibody raised against C3.

Specifications

Antigen C3
Applications ELISA, Immunodiffusion, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Goat polyclonal antibody raised against C3.
Dilution Western Blot (1:100-1:500) ELISA (1:400-1:2000) The optimal working dilution should be determined by the end user.
Formulation In 20mM sodium phosphate buffer, 140mM NaCl, pH 7.3 (0.05% sodium azide)
Gene C3
Gene Symbols C3
Host Species Goat
Immunogen Rat C3.
Quantity 2 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24232
Test Specificity Cross-reactivity to other species may exist. Antibody activity to other serum proteins is not present.
Target Species Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.