Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C4A/C4B (Turbidimetric Grade), Goat anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89116878 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-878 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-878 Supplier Abnova Corporation Supplier No. PAB13928
Add to Cart
Add to Cart

Goat polyclonal antibody raised against C4A/C4B.

Specifications

Antigen C4A/C4B
Applications Immunoassay
Classification Polyclonal
Conjugate Unconjugated
Description Goat polyclonal antibody raised against C4A/C4B.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 50mM Tris, 500mM NaCl, pH 7.5 (0.05% sodium azide)
Gene Alias C4/C4A2/C4A3/C4A4/C4A6/C4S/CO4/CPAMD2/MGC164979/RG
Gene Symbols C4A
Host Species Goat
Immunogen Purified human C4A/C4B.
Quantity 10 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 720/721
Test Specificity Monospecific by Immunoelectrophoresis (IEP) against normal human plasma and 2X concentrated normal human serum.
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.