Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CACNA1G, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89118922 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-118-922 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-118-922 Supplier Abnova Corporation Supplier No. PAB17257
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of CACNA1G.

Voltage-activated calcium channels can be distinguished based on their voltage-dependence, deactivation, and single-channel conductance. See MIM 601011. Low-voltage-activated calcium channels are referred to as 'T' type because their currents are both transient, owing to fast inactivation, and tiny, owing to small conductance. T-type channels are thought to be involved in pacemaker activity, low-threshold calcium spikes, neuronal oscillations and resonance, and rebound burst firing.[supplied by OMIM

Sequence: C-AEEISKREDASGQLS

Specifications

Antigen CACNA1G
Applications ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of CACNA1G.
Dilution ELISA (1:32000) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene CACNA1G
Gene Accession No. NP_061496.2;NP_938192.1;NP_938194.1;NP_938198.1;NP_938199.1;NP_938200.1;NP_938407.1
Gene Alias Ca(V)T.1/Cav3.1/MGC117234/NBR13
Gene Symbols CACNA1G
Host Species Goat
Immunogen A synthetic peptide corresponding to amino acids at internal region of human CACNA1G.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8913
Test Specificity This antibody is expected to recognize isoform 1 (NP_061496.2), 4 (NP_938199.1), 5 (NP_938194.1), 8 (NP_938198.1), 9 (NP_938192.1), 10 (NP_938200.1) and isoform 15 (NP_938407.1).
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.