Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CAMK2A, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89116958 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-958 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-958 Supplier Abnova Corporation Supplier No. PAB14145
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of CAMK2A.

The product of this gene belongs to the serine/threonine protein kinases family, and to the Ca(2+)/calmodulin-dependent protein kinases subfamily. Calcium signaling is crucial for several aspects of plasticity at glutamatergic synapses. This calcium calmodulin-dependent protein kinase is composed of four different chains: alpha, beta, gamma, and delta. The alpha chain encoded by this gene is required for hippocampal long-term potentiation (LTP) and spatial learning. In addition to its calcium-calmodulin (CaM)-dependent activity, this protein can undergo autophosphorylation, resulting in CaM-independent activity. Two transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq]

Sequence: C-PRTAQSEETRVWHR

Specifications

Antigen CAMK2A
Applications ELISA, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of CAMK2A.
Dilution ELISA (1:64000) Western Blot (0.1-0.3 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (3-6 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene CAMK2A
Gene Accession No. NP_057065.2;NP_741960.1
Gene Alias CAMKA/KIAA0968
Gene Symbols CAMK2A
Host Species Goat
Immunogen A synthetic peptide corresponding to internal region of human CAMK2A.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 815
Test Specificity Approx 55 and 65 KDa bands observed in human brain (cerebellum) and mouse brain lysates (calculated MW of 55.3 KDa according to human NP_057065.2 and of 54.1 KDa according to human NP_741960.1 and mouse NP_803126.1).
Target Species Human, Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.