Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CAMK2N2, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89115139 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-139 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-139 Supplier Abnova Corporation Supplier No. PAB11507

Goat polyclonal antibody raised against synthetic peptide of CAMK2N2.

This gene encodes a protein that is highly similar to the rat CaM-KII inhibitory protein, an inhibitor of calcium/calmodulin-dependent protein kinase II (CAMKII). CAMKII regulates numerous physiological functions, including neuronal synaptic plasticity through the phosphorylation of alpha-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid-type glutamate (AMPA) receptors. Studies of the similar protein in rat suggest that this protein may function as a negative regulator of CaM-KII and may act to inhibit the phosphorylation of AMPA receptors. [provided by RefSeq

Sequence: GRFGADPEGSDLS-C

Specifications

Antigen CAMK2N2
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Description calcium/calmodulin-dependent protein kinase II inhibitor 2
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene CAMK2N2
Gene Accession No. NP_150284.1
Gene Alias CAM-KIIN, CAMKIIN
Gene Symbols CAMK2N2
Host Species Goat
Immunogen A synthetic peptide corresponding to human CAMK2N2.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 94032
Content And Storage Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.