Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CAPZB, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89132092 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-132-092 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-132-092 Supplier Abnova Corporation Supplier No. PAB6046
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of CAPZB.

CAPZB is a member of the F-actin capping protein family. This gene encodes the beta subunit of the barbed-end actin binding protein. The protein regulates growth of the actin filament by capping the barbed end of growing actin filaments. [provided by RefSeq

Sequence: NDLVEALKRKQQC

Specifications

Antigen CAPZB
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of CAPZB.
Dilution ELISA (1:32000) Western Blot (0.5-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene CAPZB
Gene Accession No. NP_004921.1
Gene Alias CAPB/CAPPB/CAPZ/MGC104401/MGC129749/MGC129750
Gene Symbols CAPZB
Host Species Goat
Immunogen A synthetic peptide corresponding to C-terminus of human CAPZB.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 832
Test Specificity This antibody is expected to be specific to the beta2 isoform of capping protein, found in non-muscle cells and should not recognize the beta1 form.
Target Species Human, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.