Learn More
CARD6, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of CARD6.
Supplier: Abnova Corporation PAB6377
Description
This gene encodes a protein that contains a caspase recruitment domain (CARD), an antiparallel six-helical bundle that mediates homotypic protein-protein interactions. The encoded protein is a microtubule-associated protein that has been shown to interact with receptor-interacting protein kinases and positively modulate signal transduction pathways converging on activation of the inducible transcription factor NF-kB. [provided by RefSeq
Sequence: ATESTPSEIIERER-CSpecifications
CARD6 | |
Polyclonal | |
Unconjugated | |
ELISA (1:32000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5-10 ug/mL) The optimal working dilution should be determined by the end user. | |
CARD6 | |
CINCIN1 | |
Goat | |
Antigen affinity purification | |
RUO | |
84674 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Immunohistochemistry (PFA fixed) | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of CARD6. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
NP_115976 | |
CARD6 | |
A synthetic peptide corresponding to human CARD6. | |
100 μg | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only