Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-CBX2, Polyclonal, Abnova™

Catalog No. 89132963 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-132-963 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-132-963 Supplier Abnova Corporation Supplier No. PAB7116

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Specifications

Antigen CBX2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene Accession No. NP_005180.1;NP_116036.1
Gene Symbols CBX2
Host Species Goat
Immunogen A synthetic peptide corresponding to human CBX2.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 84733
Target Species Human
Content And Storage Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.