Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL20, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89118933 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-118-933 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-118-933 Supplier Abnova Corporation Supplier No. PAB17268
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of CCL20.

Sequence: CLGYTDRILHPK

Specifications

Antigen CCL20
Applications ELISA, Immunohistochemistry (PFA fixed)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of CCL20.
Dilution ELISA (1:16000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (6-8 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene CCL20
Gene Accession No. NP_004582.1;NP_001123518.1
Gene Alias CKb4/LARC/MIP-3a/MIP3A/SCYA20/ST38
Gene Symbols CCL20
Host Species Goat
Immunogen A synthetic peptide corresponding to internal region of human CCL20.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6364
Test Specificity This antibody is expected to recognize both isoforms (NP_004582.1; NP_001123518.1).
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.