Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDH11 Goat anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89120595 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-120-595 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-120-595 Supplier Abnova Corporation Supplier No. PAB19040
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of CDH11.

This gene encodes a type II classical cadherin from the cadherin superfamily, integral membrane proteins that mediate calcium-dependent cell-cell adhesion. Mature cadherin proteins are composed of a large N-terminal extracellular domain, a single membrane-spanning domain, and a small, highly conserved C-terminal cytoplasmic domain. Type II (atypical) cadherins are defined based on their lack of a HAV cell adhesion recognition sequence specific to type I cadherins. Expression of this particular cadherin in osteoblastic cell lines, and its upregulation during differentiation, suggests a specific function in bone development and maintenance. [provided by RefSeq

Sequence: C-ERRGHLRPSFHG

Specifications

Antigen CDH11
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of CDH11.
Dilution ELISA (1:2000) Western Blo (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene CDH11
Gene Accession No. NP_001788.2
Gene Alias CAD11/CDHOB/OB/OSF-4
Gene Symbols CDH11
Host Species Goat
Immunogen A synthetic peptide corresponding to amino acids 26-37 at internal region of human CDH11.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1009
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.