Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDKAL1, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89117684 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-684 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-684 Supplier Abnova Corporation Supplier No. PAB15614
Add to Cart
Add to Cart

Goat polyclonal antibody raised aganist synthetic peptide of CDKAL1.

Sequence: C-QVKKQRTKDLSR

Specifications

Antigen CDKAL1
Applications ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised aganist synthetic peptide of CDKAL1.
Dilution ELISA (1:64000) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene CDKAL1
Gene Accession No. NP_060244.2
Gene Alias FLJ20342/FLJ46705/MGC75469
Gene Symbols CDKAL1
Host Species Goat
Immunogen A synthetic peptide corresponding to amino acids at internal region of human CDKAL1.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54901
Test Specificity Preliminary experiments gave an approximately 39 KDa band in human brain (cerebellum) lysates after 0.5 ug/mL antibody staining. Please note that currently we cannot find an explanation in the literature for the band we observe given the calculated size of 65.1 KDa according to NP_060244.2. The 39 KDa band was successfully blocked by incubation with the immunizing peptide.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.