Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cebpz Goat anti-Chicken, Dog, Mouse, Pig, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89129169 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-129-169 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-129-169 Supplier Abnova Corporation Supplier No. PAB27655
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of Cebpz.

Sequence: RSQLDDHPESDEEN

Specifications

Antigen Cebpz
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of Cebpz.
Dilution ELISA (1:128000) Western Blot (1-3ug/ml) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA)
Gene Cebpz
Gene Accession No. NP_001019977.1
Gene Alias AI848081/CBF2/Cbf/Cebpa-rs1/MGC36087
Gene Symbols Cebpz
Host Species Goat
Immunogen A synthetic peptide corresponding to amino acids 620-633 at internal region of mouse Cebpz.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12607
Target Species Canine, Chicken, Mouse, Pig, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.