Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CECR2, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89132323 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-132-323 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-132-323 Supplier Abnova Corporation Supplier No. PAB6316
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of CECR2.

Sequence: C-PPHKPPTLPLDQS

Specifications

Antigen CECR2
Applications ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of CECR2.
Dilution ELISA (1:32000) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene CECR2
Gene Accession No. AAK15343.1
Gene Alias KIAA1740
Gene Symbols CECR2
Host Species Goat
Immunogen A synthetic peptide corresponding to C-terminus of human CECR2.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27443
Test Specificity The peptide sequence used (from AAK15343.1) differs by one residue from the RefSeq entry (XP_931107.2). It is not known whether this difference is due to a sequencing error or polymophism but this antibody is expected to recognize both sequences.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.