Learn More
CEP290 Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of CEP290.
Supplier: Abnova Corporation PAB7530
Description
This gene encodes a protein with 13 putative coiled-coil domains, a region with homology to SMC chromosome segregation ATPases, six KID motifs, three tropomyosin homology domains and an ATP/GTP binding site motif A. The protein is localized to the centrosome and cilia and has sites for N-glycosylation, tyrosine sulfation, phosphorylation, N-myristoylation, and amidation. Mutations in this gene have been associated with Joubert syndrome and nephronophthisis and the presence of antibodies against this protein is associated with several forms of cancer. [provided by RefSeq
Sequence: C-QSGAESTIPDADQSpecifications
CEP290 | |
Polyclonal | |
Unconjugated | |
ELISA (1:8000) The optimal working dilution should be determined by the end user. | |
CEP290 | |
3H11Ag/BBS14/FLJ13615/FLJ21979/JBTS5/JBTS6/KIAA0373/LCA10/MKS4/NPHP6/SLSN6/rd16 | |
Goat | |
Antigen affinity purification | |
RUO | |
80184 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of CEP290. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
NP_079390.3 | |
CEP290 | |
A synthetic peptide corresponding to human CEP290. | |
100 μg | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only