Learn More
CFH, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of CFH.
Supplier: Abnova Corporation PAB6830
Description
This gene is a member of the Regulator of Complement Activation (RCA) gene cluster and encodes a protein with twenty short concensus repeat (SCR) domains. This protein is secreted into the bloodstream and has an essential role in the regulation of complement activation, restricting this innate defense mechanism to microbial infections. Mutations in this gene have been associated with hemolytic-uremic syndrome (HUS) and chronic hypocomplementemic nephropathy. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq
Sequence: C-HLVPDRKKDQYKSpecifications
| CFH | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:32000) Western Blot (0.03-0.1 ug/mL) The optimal working dilution should be determined by the end user. | |
| CFH | |
| ARMD4/ARMS1/CFHL3/FH/FHL1/HF/HF1/HF2/HUS/MGC88246 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 3075 | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of CFH. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_000177.2 | |
| CFH | |
| A synthetic peptide corresponding to human CFH. | |
| 100 μg | |
| Primary | |
| This antibody is expected to recognize isoform a (NP_000177.2) only. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only