Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CGA Goat anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89133518 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-133-518 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-133-518 Supplier Abnova Corporation Supplier No. PAB7942
Add to Cart
Add to Cart

Goat polyclonal antibody raised against native CGA.

The four human glycoprotein hormones chorionic gonadotropin (CG), luteinizing hormone (LH), follicle stimulating hormone (FSH), and thyroid stimulating hormone (TSH) are dimers consisting of alpha and beta subunits that are associated noncovalently. The alpha subunits of these hormones are identical, however, their beta chains are unique and confer biological specificity. The protein encoded by this gene is the alpha subunit and belongs to the glycoprotein hormones alpha chain family. [provided by RefSeq

Specifications

Antigen CGA
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Description Goat polyclonal antibody raised against native CGA.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 100mM potassium phosphate buffer, 0.85% NaCl, pH 7.8 (0.05% sodium azide)
Gene CGA
Gene Alias CG-ALPHA/FSHA/GPHA1/GPHa/HCG/LHA/TSHA
Gene Symbols CGA
Host Species Goat
Immunogen Native purified human CGA.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1081
Test Specificity This antibody reacts with CGA.
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.