Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Chicken IgA Heavy Chain Secondary Antibody, Alkaline Phosphatase, Novus Biologicals™
SDP

Catalog No. NB7282 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB7282 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB7282 Supplier Novus Biologicals Supplier No. NB7282

Goat Polyclonal Antibody

IgA Heavy Chain Polyclonal antibody specifically detects IgA Heavy Chain in Chicken samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).

Specifications

Antigen IgA Heavy Chain
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Alkaline Phosphatase (AP)
Dilution Western Blot 1:100-1000, ELISA 1:500 to 1:5 000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
Gene Alias Immunoglobulin A
Gene Symbols IGHA1
Host Species Goat
Immunogen Chicken IgA
Purification Method Affinity Purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 3493
Test Specificity By immunoelectrophoresis and ELISA this reacts specifically with chicken IgA. This may cross react with IgA from other species.
Target Species Chicken
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.