Learn More
CHRNB1, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of CHRNB1.
Supplier: Abnova Corporation PAB11510
Description
The muscle acetylcholine receptor is composed of five subunits: two alpha subunits and one beta, one gamma, and one delta subunit. This gene encodes the beta subunit of the acetylcholine receptor. The acetylcholine receptor changes conformation upon acetylcholine binding leading to the opening of an ion-conducting channel across the plasma membrane. Mutations in this gene are associated with slow-channel congenital myasthenic syndrome. [provided by RefSeq
Sequence: C-QEQEDHDALKEDSpecifications
CHRNB1 | |
Polyclonal | |
Unconjugated | |
ELISA (1:1000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user. | |
CHRNB1 | |
ACHRB/CHRNB/CMS1D/CMS2A/SCCMS | |
Goat | |
Antigen affinity purification | |
RUO | |
1140 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of CHRNB1. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
NP_000738.2 | |
CHRNB1 | |
A synthetic peptide corresponding to human CHRNB1. | |
100 μg | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only