Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CKM Goat anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89116879 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-879 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-879 Supplier Abnova Corporation Supplier No. PAB13929
Add to Cart
Add to Cart

Goat polyclonal antibody raised against native CKM.

The protein encoded by this gene is a cytoplasmic enzyme involved in energy homeostasis and is an important serum marker for myocardial infarction. The encoded protein reversibly catalyzes the transfer of phosphate between ATP and various phosphogens such as creatine phosphate. It acts as a homodimer in striated muscle as well as in other tissues, and as a heterodimer with a similar brain isozyme in heart. The encoded protein is a member of the ATP:guanido phosphotransferase protein family. [provided by RefSeq

Specifications

Antigen CKM
Applications ELISA, Lateral Flow
Classification Polyclonal
Conjugate Unconjugated
Description Goat polyclonal antibody raised against native CKM.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 10mM sodium phosphate buffer, 0.85% NaCl, pH 7.2 (0.05% sodium azide)
Gene CKM
Gene Alias CKMM/M-CK
Gene Symbols CKM
Host Species Goat
Immunogen Native purified CKM from human skeletal muscle.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1158
Test Specificity human CK-MM.
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.