Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLTA/CLTB, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89115066 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-066 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-066 Supplier Abnova Corporation Supplier No. PAB11423
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of CLTA/CLTB.

Sequence: CDFNPKSSKQAKD

Specifications

Antigen CLTA/CLTB
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of CLTA/CLTB.
Dilution ELISA (1:32000) Western Blot (0.1-0.3 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene Accession No. NP_001824.1 (Gene ID : 1211);NP_009027.1 (Gene ID : 1211);NP_001070145.1 (Gene ID : 1211);NP_001825.1 (Gene ID : 1212);NP_009028.1 (Gene ID : 1212)
Gene Alias LCA
Gene Symbols CLTA
Host Species Goat
Immunogen A synthetic peptide corresponding to human CLTA/CLTB.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1211/1212
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.