Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clu Goat anti-Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89126333 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-126-333 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-126-333 Supplier Abnova Corporation Supplier No. PAB24619
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of Clu.

Sequence: C-STNNPAQANLRQE

Specifications

Antigen Clu
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of Clu.
Dilution ELISA (1:8000) Western Blot (0.1-0.3 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 0.5mg/mL Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA)
Gene Clu
Gene Accession No. NP_038520.2
Gene Alias AI893575/ApoJ/Cli/D14Ucla3/SP-40/Sgp-2/Sgp2/Sugp-2
Gene Symbols Clu
Host Species Goat
Immunogen A synthetic peptide corresponding to amino acids 312-325 at internal region of mouse Clu.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12759
Target Species Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.