Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLU Goat anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89134538 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-134-538 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-134-538 Supplier Abnova Corporation Supplier No. PAB9973
Add to Cart
Add to Cart

Goat polyclonal antibody raised against native CLU.

Specifications

Antigen CLU
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Goat polyclonal antibody raised against native CLU.
Dilution ELISA (1:5000-1:10000) Western Blot (1:5000-1:10000) Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user.
Formulation In 100mM Na2B4O7, 75mM NaCl, 5mM EDTA, pH 8.0 (0.01% sodium azide)
Gene CLU
Gene Alias AAG4/APOJ/CLI/KUB1/MGC24903/SGP-2/SGP2/SP-40/TRPM-2/TRPM2
Gene Symbols CLU
Host Species Goat
Immunogen Native purified human CLU.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1191
Test Specificity This antibody reacts with human apoLipoprotein J and has negligible cross-reactivity with Type A-I, A-II, B, C-I, C-II, C-III and E apoLipoproteins. Non-specific cross reaction of anti-apoLipoprotein antibodies with other human serum proteins is negligible.
Target Species Human
Content And Storage Store at 4°C. For long term storage store with 50% glycerol at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.