Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COL11A1, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89132986 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-132-986 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-132-986 Supplier Abnova Corporation Supplier No. PAB7140

Goat polyclonal antibody raised against synthetic peptide of COL11A1.

This gene encodes one of the two alpha chains of type XI collagen, a minor fibrillar collagen. Type XI collagen is a heterotrimer but the third alpha chain is a post-translationally modified alpha 1 type II chain. Mutations in this gene are associated with type II Stickler syndrome and with Marshall syndrome. A single-nucleotide polymorphism in this gene is also associated with susceptibility to lumbar disc herniation. Three transcript variants encoding different isoforms have been identified for this gene. [provided by RefSeq

Sequence: C-EDYDSQRKNSEDTLY

Specifications

Antigen COL11A1
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Description collagen, type XI, alpha 1
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene COL11A1
Gene Accession No. NP_001845.3;NP_542196.2;NP_542197.2
Gene Alias CO11A1, COLL6, STL2
Gene Symbols COL11A1
Host Species Goat
Immunogen A synthetic peptide corresponding to human COL11A1.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1301
Test Specificity This antibody is expected to recognize all three reported isoforms (NP_001845.3, NP_542196.2 and NP_542197.2).
Content And Storage Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.