Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-COL2A1, Polyclonal, Abnova™

Catalog No. 89116876 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-876 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-876 Supplier Abnova Corporation Supplier No. PAB13923

Specifications

Antigen COL2A1
Applications Dot Blot, ELISA, Immunocytochemistry, Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate Unconjugated
Formulation In 100mM BBS, pH 8.0
Gene Symbols COL2A1
Host Species Goat
Immunogen Bovine COL2A1.
Quantity 200 μg
Primary or Secondary Primary
Gene ID (Entrez) 407142
Target Species Bovine
Content And Storage Store at 4°C.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.