Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COL3A1, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89117016 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-016 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-016 Supplier Abnova Corporation Supplier No. PAB14346
Add to Cart
Add to Cart

Goat polyclonal antibody raised against native COL3A1.

This gene encodes the pro-alpha1 chains of type III collagen, a fibrillar collagen that is found in extensible connective tissues such as skin, lung, uterus, intestine and the vascular system, frequently in association with type I collagen. Mutations in this gene are associated with Ehlers-Danlos syndrome types IV, and with aortic and arterial aneurysms. Two transcripts, resulting from the use of alternate polyadenylation signals, have been identified for this gene. [provided by R. Dalgleish

Specifications

Antigen COL3A1
Applications Dot Blot, ELISA, Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Goat polyclonal antibody raised against native COL3A1.
Dilution Immunocytochemistry (1:10-1:20) Immunohistochemistry (1:10-1:20) The optimal working dilution should be determined by the end user.
Formulation In 100mM BBS, pH 8.2
Gene COL3A1
Gene Alias EDS4A/FLJ34534
Gene Symbols COL3A1
Host Species Goat
Immunogen Native human COL3A1.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1281
Test Specificity Reacts with conformational determinants on human type III collagen as demonstrated by ELISA. May react with type III collagen from other species. Exhibits <10% cross reactivity with collagen type I, II, IV, V, VI. This antibody has not been tested for reactivity with other ECM proteins (e.g., laminin, fibronectin).
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.