Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COL4, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89137921 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-921 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-921 Supplier Abnova Corporation Supplier No. PAB14347
Add to Cart
Add to Cart

Goat polyclonal antibody raised against native COL4.

Specifications

Antigen COL4
Applications Dot Blot, ELISA, Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Goat polyclonal antibody raised against native COL4.
Dilution Immunocytochemistry (1:10-1:20) Immunohistochemistry (1:10-1:20) The optimal working dilution should be determined by the end user.
Formulation In 100mM BBS, pH 8.2
Host Species Goat
Immunogen Native human COL4.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Reacts with conformational determinants on human type IV collagen as demonstrated by ELISA. May react with type IV collagen from other species. Exhibits <10% cross reactivity with collagen type I, II, III, V, VI. This antibody has not been tested for reactivity with other ECM proteins (e.g., laminin, fibronectin).
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.