Learn More
COL4A3BP Goat anti-Mouse, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of COL4A3BP.
Supplier: Abnova Corporation PAB7393
Description
This gene encodes a kinase that specifically phosphorylates the N-terminal region of the noncollagenous domain of the alpha 3 chain of type IV collagen, known as the Goodpasture antigen. Goodpasture disease is the result of an autoimmune response directed at this antigen. One isoform of this protein is also involved in ceramide intracellular transport. Three transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: DHDSAPLNNRCSpecifications
COL4A3BP | |
Polyclonal | |
Unconjugated | |
ELISA (1:32000) Western Blot (0.5-1.5 ug/mL) The optimal working dilution should be determined by the end user. | |
COL4A3BP | |
CERT/CERTL/FLJ20597/GPBP/STARD11 | |
Goat | |
Antigen affinity purification | |
RUO | |
10087 | |
Human | |
Liquid |
ELISA, Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of COL4A3BP. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
NP_005704.1;NP_112729.1 | |
COL4A3BP | |
A synthetic peptide corresponding to human COL4A3BP. | |
100 μg | |
Primary | |
This antibody is expected to recognize both reported isoforms (NP_005704.1 and NP_112729.1). | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only