Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Collagen Type IV, Biotin, Polyclonal, Abnova™

Catalog No. 89137859 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-859 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-859 Supplier Abnova Corporation Supplier No. PAB14220

Specifications

Antigen Collagen Type IV
Applications ELISA, Immunocytochemistry
Classification Polyclonal
Conjugate Biotin
Formulation In 100mM BBS, pH 8.0 (0.09% sodium azide)
Host Species Goat
Immunogen Native purified human and bovine type IV collagen.
Quantity 200 μg
Primary or Secondary Primary
Target Species Bovine, Human
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.