Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CP, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89117602 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-602 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-602 Supplier Abnova Corporation Supplier No. PAB15405
Add to Cart
Add to Cart

Goat polyclonal antibody raised against CP.

The protein encoded by this gene is a metalloprotein that binds most of the copper in plasma and is involved in the peroxidation of Fe(II)transferrin to Fe(III) transferrin. Mutations in this gene cause aceruloplasminemia, which results in iron accumulation and tissue damage, and is associated with diabetes and neurologic abnormalities. [provided by RefSeq

Specifications

Antigen CP
Applications ELISA, Immunodiffusion, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Goat polyclonal antibody raised against CP.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 20mM sodium phosphate buffer, 140mM NaCl, pH 7.3 (0.05% sodium azide)
Gene CP
Gene Alias CP-2
Gene Symbols CP
Host Species Goat
Immunogen Human CP.
Quantity 2 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1356
Test Specificity Cross-reactivity to other species may exist. Antibody activity to other serum proteins is not present.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.